Gene Transcripts

Gene Symbol
RELL2
Entrez Gene ID
285613
Gene Name
RELT like 2
Gene Aliases
C5orf16
Chromosome Location
Chr. 5: 141636946 - 141641077 on Build GRCh38
UniGene ID
Hs.120094
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130029.1 NP_001123501.1 3 1053
NM_173828.4 NP_776189.3 2 1481
XM_005268414.3 XP_005268471.1 3 1057
XM_011537624.1 XP_011535926.1 3 1073
XM_011537625.2 XP_011535927.1 2 1485
GenBank mRNA AK054889.1 - 2 1481
AK075064.1 - 2 308
AY358163.1 - 2 1417
BC063469.1 - 3 522 5211730

View Full Product Details