Gene Transcripts

Gene Symbol
ENHO
Entrez Gene ID
375704
Gene Name
energy homeostasis associated
Gene Aliases
C9orf165, UNQ470
Chromosome Location
Chr. 9: 34521042 - 34523039 on Build GRCh38
UniGene ID
Hs.522085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198573.2 NP_940975.2 2 933
GenBank mRNA AY358658.1 - 1 661
BC022101.1 - 3 743 4871461
BQ932712.1 - 2 783
BX111579.1 - 1 520

View Full Product Details