Gene Transcripts

Gene Symbol
ATP6AP2
Entrez Gene ID
10159
Gene Name
ATPase H+ transporting accessory protein 2
Gene Aliases
APT6M8-9, ATP6IP2, ATP6M8-9, ELDF10, HT028, M8-9, MRXE, MRXSH, MSTP009, PRR, RENR, XMRE, XPDS
Chromosome Location
Chr. X: 40580964 - 40606637 on Build GRCh38
UniGene ID
Hs.495960
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005765.2 NP_005756.2 4, 5 508
GenBank mRNA AF109363.1 - 4, 5 469
AF248966.1 - 4, 5 513
AF291814.1 - 4, 5 495
AK075382.1 - 4, 5 566
AK296614.1 - 3, 4 371
AK314633.1 - 4, 5 459
AL049929.1 - 3, 4 348
AY429341.1 - 3, 4 371
BC084541.1 - 4, 5 488 6575732

View Full Product Details