Gene Transcripts

Gene Symbol
CBWD5
Entrez Gene ID
220869
Gene Name
COBW domain containing 5
Gene Aliases
DC36
Chromosome Location
Chr. 9: 65666374 - 65734041 on Build GRCh38
UniGene ID
Hs.355950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286835.1 NP_001273764.1 9 883
XM_005272748.1 XP_005272805.1 8 755
XM_005272749.1 XP_005272806.1 8 755
XM_005272750.1 XP_005272807.1 7 695
XM_006716914.1 XP_006716977.1 8 755
XM_017014456.1 XP_016869945.1 9 686
XM_017014458.1 XP_016869947.1 8 755
XM_017014459.1 XP_016869948.1 10 934
GenBank mRNA AK097639.1 - 7 1071
AK292585.1 - 9 823
BC067803.1 - 9 821 5294979
BC141856.1 - 9 821
CD244615.1 - 1 62

Gene Symbol
CBWD1
Entrez Gene ID
55871
Gene Name
COBW domain containing 1
Gene Aliases
COBP
Chromosome Location
Chr. 9: 121038 - 179083 on Build GRCh38
UniGene ID
Hs.531330
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145355.1 NP_001138827.1 9 856
NM_001145356.1 NP_001138828.1 8 758
NM_018491.3 NP_060961.3 8 758
XM_005251513.4 XP_005251570.1 7 706
XM_005251517.4 XP_005251574.1 8 758
XM_011517961.2 XP_011516263.1 9 850
XM_011517962.2 XP_011516264.1 8 758
XM_011517963.1 XP_011516265.1 5 296
XM_017014925.1 XP_016870414.1 8 758
GenBank mRNA AF212253.1 - 8 726
AF257330.2 - 8 716
AK290648.1 - 9 828
AK297292.1 - 9 797
AK315855.1 - 8 682
AY343911.1 - 8 715
BC005996.1 - 8 758 4069589
BC009573.1 - 9 809 4133536
BC013432.1 - 8 729 4133722
BC132759.1 - 8 692 40146781

Gene Symbol
CBWD3
Entrez Gene ID
445571
Gene Name
COBW domain containing 3
Gene Aliases
bA561O23.1
Chromosome Location
Chr. 9: 68232437 - 68300035 on Build GRCh38
UniGene ID
Hs.645337
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291821.1 NP_001278750.1 7 679
NM_201453.3 NP_958861.2 8 739
XM_005277637.2 XP_005277694.1 8 818
XM_006717116.1 XP_006717179.1 8 818
XM_011518706.2 XP_011517008.1 3 269
XM_017014731.1 XP_016870220.1 8 3190
XM_017014733.1 XP_016870222.1 3 207
GenBank mRNA AK297912.1 - 7 641
AY344633.1 - 8 715

View Full Product Details