Gene Transcripts
- Gene Symbol
- SMA4
- Entrez Gene ID
- 11039
- Gene Name
- glucuronidase beta pseudogene
- Gene Aliases
- SMA3, b55C20.2
- Chromosome Location
- Chr. 5: 70127462 - 70290177 on Build GRCh38
- UniGene ID
- Hs.582500
- Species
- Homo sapiens
- Gene Symbol
- GUSBP1
- Entrez Gene ID
- 728411
- Gene Name
- glucuronidase, beta pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 5: 21459480 - 21589372 on Build GRCh38
- UniGene ID
- Hs.88181
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027028.2 |
|
6 |
693 |
|
GenBank mRNA |
BC147000.1 |
- |
6 |
647 |
|
- Gene Symbol
- GUSBP3
- Entrez Gene ID
- 653188
- Gene Name
- glucuronidase, beta pseudogene 3
- Gene Aliases
- Chromosome Location
- Chr. 5: 69639463 - 69710527 on Build GRCh38
- UniGene ID
- Hs.631974
- Species
- Homo sapiens
- Gene Symbol
- GUSBP4
- Entrez Gene ID
- 375513
- Gene Name
- glucuronidase, beta pseudogene 4
- Gene Aliases
- C6orf216, GUSBL2, SMA3-L, SMAC3L
- Chromosome Location
- Chr. 6: 57908553 - 57936929 on Build GRCh38
- UniGene ID
- Hs.561539
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_132999.1 |
|
4 |
481 |
|
NR_133000.1 |
|
4 |
481 |
|
GenBank mRNA |
AW955360.1 |
- |
3 |
309 |
|
- Gene Symbol
- LINC00680-GUSBP4
- Entrez Gene ID
- 106660613
- Gene Name
- LINC00680-GUSBP4 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 6: 57919881 - 57961446 on Build GRCh38
- UniGene ID
- Hs.561539
- Species
- Homo sapiens
- Gene Symbol
- SMA5
- Entrez Gene ID
- 11042
- Gene Name
- glucuronidase beta pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 5: 70481043 - 70585722 on Build GRCh38
- UniGene ID
- Hs.529793
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034021.1 |
|
7 |
806 |
|
GenBank mRNA |
CN312642.1 |
- |
4 |
596 |
|
X83301.1 |
- |
7 |
806 |
|
View Full Product Details