Gene Transcripts

Gene Symbol
NME6
Entrez Gene ID
10201
Gene Name
NME/NM23 nucleoside diphosphate kinase 6
Gene Aliases
IPIA-ALPHA, NDK 6, NM23-H6
Chromosome Location
Chr. 3: 48289369 - 48302259 on Build GRCh38
UniGene ID
Hs.465558
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308426.1 NP_001295355.1 3 467
NM_001308427.1 NP_001295356.1 3 276
NM_001308428.1 NP_001295357.1 3 267
NM_001308430.1 NP_001295359.1 3 250
NM_001308431.1 NP_001295360.1 3 467
NM_001308433.1 NP_001295362.1 3 267
NM_001308434.1 NP_001295363.1 2 370
NM_001308435.1 NP_001295364.1 3 467
NM_005793.4 NP_005784.1 3 250
XM_017005511.1 XP_016861000.1 3 178
XM_017005513.1 XP_016861002.1 3 192
XM_017005514.1 XP_016861003.1 4 449
XM_017005515.1 XP_016861004.1 3 245
XM_017005516.1 XP_016861005.1 3 207
XM_017005518.1 XP_016861007.1 1 18
GenBank mRNA AF051941.1 - 3 183
AK222555.1 - 2 149
AK294809.1 - 3 223
AK297364.1 - 3 176
AK311152.1 - 1 18
BC001808.2 - 3 197 2966629
BC012828.2 - 3 197 2966629
BE866839.1 - 3 218
BG284082.1 - 3 196
BG530435.1 - 2 72
CA848473.1 - 3 246
CD701581.1 - 3 220
CT002144.1 - 3 467
DA398008.1 - 3 223
HY090220.1 - 3 220
U90449.1 - 3 197

View Full Product Details