Gene Transcripts

Gene Symbol
Nlrp1a
Entrez Gene ID
195046
Gene Name
NLR family, pyrin domain containing 1A
Gene Aliases
CARD7, DEFCAP, Gm14, Gm15, NAC, Nalp1, Nalp1a, Nlrp1, PP1044
Chromosome Location
Chr. 11: 71091197 - 71144704 on Build GRCm38
UniGene ID
Mm.240227
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004142.2 NP_001004142.2 11 3270
GenBank mRNA AY355339.1 - 12 3355
DQ117601.1 - 10 3211
KC539856.1 - 10 3052
KC539857.1 - 10 3052
KC539858.1 - 10 3052
KC539859.1 - 10 3052
KC539860.1 - 10 3052
KC539861.1 - 10 3052

View Full Product Details