Gene Transcripts

Gene Symbol
Sprr2h
Entrez Gene ID
20762
Gene Name
small proline-rich protein 2H
Gene Aliases
AI661967
Chromosome Location
Chr. 3: 92385685 - 92387324 on Build GRCm38
UniGene ID
Mm.10693
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011474.3 NP_035604.1 2 567
GenBank mRNA AJ005566.1 - 1 539
AY158992.1 - 2 567

View Full Product Details