Gene Transcripts

Gene Symbol
Trav7-4
Entrez Gene ID
386450
Gene Name
T cell receptor alpha variable 7-4
Gene Aliases
Gm13949, OTTMUSG00000015161
Chromosome Location
Chr. 14: 53461209 - 53461738 on Build GRCm38
UniGene ID
Mm.425889
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA X63928.1 - 48

View Full Product Details