Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006108.3 | NP_006099.2 | 4 | 761 | |
GenBank mRNA | AB018305.2 | - | 3 | 246 | |
AB051390.1 | - | 4 | 517 | ||
AK074803.1 | - | 4 | 707 | ||
AK291780.1 | - | 4 | 709 | ||
BC136513.1 | - | 4 | 703 | 9020503 | |
BC136563.1 | - | 4 | 703 | 9020553 |