Gene Transcripts
- Gene Symbol
- Anapc4
- Entrez Gene ID
- 52206
- Gene Name
- anaphase promoting complex subunit 4
- Gene Aliases
- 2610306D21Rik, APC4, D5Ertd249e
- Chromosome Location
- Chr. 5: 52834126 - 52866742 on Build GRCm38
- UniGene ID
- Mm.272568
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024213.2 |
NP_077175.1
|
7, 8 |
735 |
|
GenBank mRNA |
AK088759.1 |
- |
7, 8 |
725 |
|
AK146000.1 |
- |
7, 8 |
732 |
|
AK148625.1 |
- |
7, 8 |
744 |
|
AK166061.1 |
- |
7, 8 |
858 |
|
AK168031.1 |
- |
7, 8 |
732 |
|
AK168897.1 |
- |
8, 9 |
817 |
|
AK192250.1 |
- |
2, 3 |
127 |
|
BC016237.1 |
- |
7, 8 |
718 |
4504779 |
BC024870.1 |
- |
7, 8 |
735 |
4949079 |
View Full Product Details