Gene Transcripts

Gene Symbol
Vmn2r77
Entrez Gene ID
546983
Gene Name
vomeronasal 2, receptor 77
Gene Aliases
EG546983
Chromosome Location
Chr. 7: 86795141 - 86812032 on Build GRCm38
UniGene ID
Mm.469904
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105188.1 NP_001098658.1 2 465

View Full Product Details