Gene Transcripts

Gene Symbol
CAP2
Entrez Gene ID
10486
Gene Name
CAP, adenylate cyclase-associated protein, 2 (yeast)
Gene Aliases
Chromosome Location
Chr. 6: 17393216 - 17557792 on Build GRCh38
UniGene ID
Hs.132902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006366.2 NP_006357.1 6 757
XM_011514233.2 XP_011512535.1 5 968
GenBank mRNA AK297538.1 - 5 473
AK308461.1 - 5 472
AK312362.1 - 6 885
BC008481.1 - 6 550 4295984
CR457022.1 - 5 514
U02390.1 - 6 597

View Full Product Details