Gene Transcripts
- Gene Symbol
- Cln6
- Entrez Gene ID
- 76524
- Gene Name
- ceroid-lipofuscinosis, neuronal 6
- Gene Aliases
- 1810065L06Rik, AW743417, D9Bwg1455e, nclf
- Chromosome Location
- Chr. 9: 62838787 - 62852002 on Build GRCm38
- UniGene ID
- Mm.283636
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033175.2 |
NP_001028347.1
|
6, 7 |
755 |
|
GenBank mRNA |
AK154414.1 |
- |
6, 7 |
756 |
|
AK154551.1 |
- |
6, 7 |
749 |
|
AK177422.1 |
- |
2, 3 |
195 |
|
AK209042.1 |
- |
2, 3 |
195 |
|
AK211299.1 |
- |
2, 3 |
195 |
|
AK216712.1 |
- |
2, 3 |
195 |
|
BC116794.1 |
- |
5, 6 |
581 |
|
BC116798.1 |
- |
5, 6 |
581 |
|
BC144722.1 |
- |
5, 6 |
640 |
|
View Full Product Details