Gene Transcripts

Gene Symbol
POSTN
Entrez Gene ID
10631
Gene Name
periostin
Gene Aliases
OSF-2, OSF2, PDLPOSTN, PN
Chromosome Location
Chr. 13: 37562582 - 37598844 on Build GRCh38
UniGene ID
Hs.136348
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135934.1 NP_001129406.1 5 622
NM_001135935.1 NP_001129407.1 5 622
NM_001135936.1 NP_001129408.1 5 622
NM_001286665.1 NP_001273594.1 5 622
NM_001286666.1 NP_001273595.1 5 622
NM_001286667.1 NP_001273596.1 5 622
NM_006475.2 NP_006466.2 5 622
XM_005266231.2 XP_005266288.1 5 617
XM_005266232.2 XP_005266289.1 5 617
XM_017020355.1 XP_016875844.1 5 617
XM_017020356.1 XP_016875845.1 5 617
GenBank mRNA AK300261.1 - 5 736
AY140646.1 - 5 515
AY918092.1 - 5 515
BC106709.1 - 5 515 40007999
BC106710.1 - 5 515 40008000
BY795054.2 - 5 529
D13665.1 - 5 531
D13666.1 - 5 515
EU262883.1 - 5 504
EU262884.1 - 5 504
EU262885.1 - 5 504
EU262886.1 - 5 504
GU354210.1 - 5 515

View Full Product Details