Gene Transcripts

Gene Symbol
IGF2BP2
Entrez Gene ID
10644
Gene Name
insulin like growth factor 2 mRNA binding protein 2
Gene Aliases
IMP-2, IMP2, VICKZ2
Chromosome Location
Chr. 3: 185643739 - 185825051 on Build GRCh38
UniGene ID
Hs.35354
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007225.1 NP_001007226.1 2 284
NM_001291869.1 NP_001278798.1 2 284
NM_006548.4 NP_006539.3 2 284
XM_017005558.1 XP_016861047.1 2 289
XM_017005559.1 XP_016861048.1 2 290
XM_017005560.1 XP_016861049.1 2 186
XM_017005561.1 XP_016861050.1 2 291
XM_017005562.1 XP_016861051.1 2 290
XM_017005563.1 XP_016861052.1 2 290
XM_017005564.1 XP_016861053.1 2 290
GenBank mRNA AF117107.1 - 2 251
AK290664.1 - 2 285
AK309611.1 - 2 238
BC021290.2 - 2 269 5090334

View Full Product Details