Gene Transcripts
- Gene Symbol
- Huwe1
- Entrez Gene ID
- 59026
- Gene Name
- HECT, UBA and WWE domain containing 1
- Gene Aliases
- 5430439H10Rik, AU041296, Arf-bp1, C430014N20Rik, C80292, Gm1718, Ib772, LASU1, Mule, Ureb1
- Chromosome Location
- Chr. X: 151800807 - 151935417 on Build GRCm38
- UniGene ID
- Mm.27372
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021523.4 |
NP_067498.4
|
8, 9 |
1103 |
|
GenBank mRNA |
AK031374.1 |
- |
9, 10 |
1130 |
|
AK049460.1 |
- |
8, 9 |
1059 |
|
AK142956.1 |
- |
9, 10 |
1090 |
|
AK163416.1 |
- |
9, 10 |
1253 |
|
AK165140.1 |
- |
8, 9 |
1039 |
|
AK165648.1 |
- |
9, 10 |
1176 |
|
AY772010.3 |
- |
8, 9 |
1040 |
|
AY929611.1 |
- |
6, 7 |
650 |
|
DQ097265.1 |
- |
8, 9 |
1059 |
|
View Full Product Details