Gene Transcripts

Gene Symbol
Mfsd10
Entrez Gene ID
68294
Gene Name
major facilitator superfamily domain containing 10
Gene Aliases
0610009O03Rik, Tetran
Chromosome Location
Chr. 5: 34633644 - 34637215 on Build GRCm38
UniGene ID
Mm.34564
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026660.2 NP_080936.1 10 1296
GenBank mRNA AK003763.1 - 5 564
AK018736.1 - 10 1309
AK089055.1 - 10 1313
AK144407.1 - 1 16
AK152438.1 - 10 1282
BC023012.1 - 10 1296 5376230
BC064775.1 - 10 1194 30023877

View Full Product Details