Gene Transcripts

Gene Symbol
IFT27
Entrez Gene ID
11020
Gene Name
intraflagellar transport 27
Gene Aliases
BBS19, RABL4, RAYL
Chromosome Location
Chr. 22: 36758202 - 36776133 on Build GRCh38
UniGene ID
Hs.415172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177701.2 NP_001171172.1 5 695
NM_006860.4 NP_006851.1 5 692
XM_006724106.2 XP_006724169.1 6 456
XM_017028540.1 XP_016884029.1 5 699
GenBank mRNA AB209424.1 - 5 687
AL022729.1 - 5 636
BC000566.2 - 5 673 3163341
BM711121.1 - 5 283
BT006815.1 - 5 269
CR456558.1 - 5 311

View Full Product Details