Gene Transcripts

Gene Symbol
LILRA2
Entrez Gene ID
11027
Gene Name
leukocyte immunoglobulin like receptor A2
Gene Aliases
CD85H, ILT1, LIR-7, LIR7
Chromosome Location
Chr. 19: 54572997 - 54587560 on Build GRCh38
UniGene ID
Hs.655593
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130917.2 NP_001124389.2 3 231
NM_001290270.1 NP_001277199.1 2 195
NM_001290271.1 NP_001277200.1 4 670
NM_006866.3 NP_006857.2 4 803
XM_006722986.1 XP_006723049.1 4 803
XM_011526385.1 XP_011524687.1 4 803
XM_011526386.1 XP_011524688.1 3 767
XM_011526387.1 XP_011524689.1 3 767
XM_011526388.1 XP_011524690.1 3 767
XM_011526390.1 XP_011524692.1 4 803
XM_011526391.1 XP_011524693.1 4 803
XM_017026224.1 XP_016881713.1 4 803
GenBank mRNA AF025531.1 - 3 222
AK130170.1 - 3 261
AK222651.1 - 3 227
AK310088.1 - 4 652
BC017412.1 - 3 233 4618777
BC027916.1 - 3 216 5223154
BP296918.1 - 3 272
EU915609.1 - 2 164
EU915610.1 - 3 200
U70665.1 - 3 232
U82275.1 - 3 270
U82276.1 - 2 185
U82277.1 - 3 183
U82278.1 - 3 183

View Full Product Details