Gene Transcripts

Gene Symbol
ADAP1
Entrez Gene ID
11033
Gene Name
ArfGAP with dual PH domains 1
Gene Aliases
CENTA1, GCS1L, p42IP4
Chromosome Location
Chr. 7: 897900 - 955407 on Build GRCh38
UniGene ID
Hs.602573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284308.1 NP_001271237.1 5 524
NM_001284309.1 NP_001271238.1 5 464
NM_001284310.1 NP_001271239.1 4 439
NM_001284311.1 NP_001271240.1 3 292
NM_006869.3 NP_006860.1 5 697
GenBank mRNA AB097049.1 - 5 671
AF082324.1 - 5 671
AJ006422.1 - 5 693
AK092471.1 - 4 441
AK124979.1 - 3 543
AK290517.1 - 6 643
AK300644.1 - 3 276
AK300859.1 - 3 279
AK300999.1 - 5 524
BC033747.1 - 5 498 5195416
BM837987.1 - 3 292
DA872042.1 - 5 464

View Full Product Details