Gene Transcripts

Gene Symbol
ADRM1
Entrez Gene ID
11047
Gene Name
adhesion regulating molecule 1
Gene Aliases
ARM-1, ARM1, GP110
Chromosome Location
Chr. 20: 62302056 - 62308862 on Build GRCh38
UniGene ID
Hs.90107
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281437.1 NP_001268366.1 3 404
NM_001281438.1 NP_001268367.1 3 408
NM_007002.3 NP_008933.2 4 525
NM_175573.2 NP_783163.1 4 521
XM_005260257.1 XP_005260314.1 5 612
XM_011528503.1 XP_011526805.1 4 1360
XM_017027602.1 XP_016883091.1 4 1360
XM_017027603.1 XP_016883092.1 5 612
XM_017027604.1 XP_016883093.1 4 1356
XM_017027605.1 XP_016883094.1 4 1360
XM_017027606.1 XP_016883095.1 3 1243
XM_017027607.1 XP_016883096.1 3 1239
XM_017027608.1 XP_016883097.1 2 1029
XM_017027609.1 XP_016883098.1 2 1029
XM_017027610.1 XP_016883099.1 2 1029
GenBank mRNA AK054985.1 - 1588
AK297567.1 - 4 419
AK307459.1 - 455
AL558522.3 - 4 457
BC010733.1 - 1 37
BC017245.2 - 4 421 4890786
BG337785.1 - 3 364
CR749213.1 - 1 2761
D64154.1 - 4 416
HY178253.1 - 3 354

View Full Product Details