Gene Transcripts

Gene Symbol
ADRM1
Entrez Gene ID
11047
Gene Name
adhesion regulating molecule 1
Gene Aliases
ARM-1, ARM1, GP110
Chromosome Location
Chr. 20: 62302056 - 62308862 on Build GRCh38
UniGene ID
Hs.90107
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281437.1 NP_001268366.1 3 397
NM_001281438.1 NP_001268367.1 3 401
NM_007002.3 NP_008933.2 4 518
NM_175573.2 NP_783163.1 4 514
XM_005260257.1 XP_005260314.1 5 605
XM_011528503.1 XP_011526805.1 4 1353
XM_017027602.1 XP_016883091.1 4 1353
XM_017027603.1 XP_016883092.1 5 605
XM_017027604.1 XP_016883093.1 4 1349
XM_017027605.1 XP_016883094.1 4 1353
XM_017027606.1 XP_016883095.1 3 1236
XM_017027607.1 XP_016883096.1 3 1232
XM_017027608.1 XP_016883097.1 2 1022
XM_017027609.1 XP_016883098.1 2 1022
XM_017027610.1 XP_016883099.1 2 1022
GenBank mRNA AK054985.1 - 1581
AK297567.1 - 4 412
AK307459.1 - 448
AL558522.3 - 4 450
BC010733.1 - 1 30
BC017245.2 - 4 414 4890786
BG337785.1 - 3 357
CR749213.1 - 1 2754
D64154.1 - 4 409
HY178253.1 - 3 347

View Full Product Details