Gene Transcripts

Gene Symbol
Cyp2c7
Entrez Gene ID
29298
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 7
Gene Aliases
Cyp2c39
Chromosome Location
Chr. 1: 147422999 - 148119979 on Build Rnor_6.0
UniGene ID
Rn.1247
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017158.2 NP_058854.1 9 1442
XM_008759609.1 XP_008757831.1 9 1564
GenBank mRNA BC097939.1 - 9 1425
BC127503.1 - 9 1481 7371672
FQ218330.1 - 8 2478
M18335.1 - 9 1443
M31031.1 - 9 1400

Gene Symbol
LOC100361547
Entrez Gene ID
100361547
Gene Name
Cytochrome P450, family 2, subfamily c, polypeptide 7-like
Gene Aliases
Chromosome Location
Chr. 1: 148232438 - 148287458 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759635.1 XP_008757857.1 9 1298

View Full Product Details