Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007030.2 | NP_008961.1 | 3 | 453 | |
XM_005248237.3 | XP_005248294.2 | 3 | 518 | ||
XM_017008993.1 | XP_016864482.1 | 3 | 2649 | ||
XM_017008994.1 | XP_016864483.1 | 5 | 504 | ||
GenBank mRNA | AB017016.1 | - | 3 | 364 | |
AK054920.1 | - | 2 | 1050 | ||
AK098392.1 | - | 3 | 409 | ||
AY319261.1 | - | 2 | 331 | ||
BC131506.1 | - | 2 | 349 | 40082752 | |
BC152382.1 | - | 2 | 334 | 100013441 | |
BT020035.1 | - | 2 | 331 | ||
DB478250.2 | - | 3 | 453 |