Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134358.1 | NP_599185.1 | 7 | 854 | |
GenBank mRNA | BC083694.1 | - | 7 | 1251 | |
BC158686.1 | - | 7 | 854 | 8374870 | |
FQ214448.1 | - | 7 | 829 | ||
FQ220404.1 | - | 7 | 807 | ||
FQ220639.1 | - | 7 | 870 | ||
FQ220740.1 | - | 7 | 855 | ||
FQ220961.1 | - | 7 | 855 | ||
FQ229287.1 | - | 7 | 857 | ||
FQ229512.1 | - | 7 | 870 | ||
FQ230125.1 | - | 7 | 807 | ||
M29295.1 | - | 6 | 707 |