Gene Transcripts

Gene Symbol
Mospd3
Entrez Gene ID
288557
Gene Name
motile sperm domain containing 3
Gene Aliases
Chromosome Location
Chr. 12: 22164983 - 22169251 on Build Rnor_6.0
UniGene ID
Rn.16636
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025629.1 NP_001020800.1 3 499
XM_006249115.2 XP_006249177.1 4 559
GenBank mRNA BC099230.1 - 3 499
FQ220320.1 - 3 598

View Full Product Details