Gene Transcripts

Gene Symbol
Gem
Entrez Gene ID
297902
Gene Name
GTP binding protein overexpressed in skeletal muscle
Gene Aliases
Chromosome Location
Chr. 5: 25339479 - 25353661 on Build Rnor_6.0
UniGene ID
Rn.40436
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106637.1 NP_001100107.1 482
GenBank mRNA BC168988.1 - 432

View Full Product Details