Gene Transcripts

Gene Symbol
B4galnt4
Entrez Gene ID
309105
Gene Name
beta-1,4-N-acetyl-galactosaminyl transferase 4
Gene Aliases
RGD1310052
Chromosome Location
Chr. 1: 213870328 - 213881233 on Build Rnor_6.0
UniGene ID
Rn.98521
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107562.1 NP_001101032.1 11 1221
GenBank mRNA FQ211564.1 - 11 1053

View Full Product Details