Gene Transcripts

Gene Symbol
Rc3h2
Entrez Gene ID
311909
Gene Name
ring finger and CCCH-type domains 2
Gene Aliases
Mnab
Chromosome Location
Chr. 3: 21621976 - 21676780 on Build Rnor_6.0
UniGene ID
Rn.134067
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107839.1 NP_001101309.1 4 443
XM_008761754.1 XP_008759976.1 4 544
XM_008761755.1 XP_008759977.1 4 544

View Full Product Details