Gene Transcripts

Gene Symbol
AKAP11
Entrez Gene ID
11215
Gene Name
A-kinase anchoring protein 11
Gene Aliases
AKAP-11, AKAP220, PPP1R44, PRKA11
Chromosome Location
Chr. 13: 42271470 - 42323267 on Build GRCh38
UniGene ID
Hs.105105
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016248.3 NP_057332.1 8 1701
XM_005266247.1 XP_005266304.1 8 1702
XM_005266248.3 XP_005266305.1 7 1595
XM_005266249.3 XP_005266306.1 8 1702
XM_005266250.3 XP_005266307.1 8 1702
XM_011534903.2 XP_011533205.1 8 1698
XM_011534904.1 XP_011533206.1 8 1894
XM_011534905.1 XP_011533207.1 7 1652
XM_011534906.2 XP_011533208.1 7 1870
XM_017020381.1 XP_016875870.1 8 1752
XM_017020382.1 XP_016875871.1 6 1542
XM_017020383.1 XP_016875872.1 6 1730
GenBank mRNA AF176555.1 - 8 1701
CR627087.1 - 7 1864

View Full Product Details