Gene Transcripts

Gene Symbol
Zcchc17
Entrez Gene ID
500555
Gene Name
zinc finger, CCHC domain containing 17
Gene Aliases
RGD1565267
Chromosome Location
Chr. 5: 148535519 - 148577312 on Build Rnor_6.0
UniGene ID
Rn.63831
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109267.1 NP_001102737.1 4, 5 373
XM_006238968.2 XP_006239030.1 4, 5 393
XM_006238969.2 XP_006239031.1 3, 4 329
XM_006238970.2 XP_006239032.1 2, 3 208

View Full Product Details