Gene Transcripts
- Gene Symbol
- DCTN3
- Entrez Gene ID
- 11258
- Gene Name
- dynactin subunit 3
- Gene Aliases
- DCTN-22, DCTN22
- Chromosome Location
- Chr. 9: 34613545 - 34620523 on Build GRCh38
- UniGene ID
- Hs.511768
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281426.1 |
NP_001268355.1
|
4, 5 |
411 |
|
NM_001281427.1 |
NP_001268356.1
|
4, 5 |
411 |
|
NM_007234.4 |
NP_009165.1
|
4, 5 |
411 |
|
NM_024348.3 |
NP_077324.1
|
4, 5 |
411 |
|
GenBank mRNA |
AF082513.1 |
- |
4, 5 |
368 |
|
AK314730.1 |
- |
4, 5 |
379 |
|
AW953496.1 |
- |
4, 5 |
356 |
|
BC000319.2 |
- |
4, 5 |
362 |
2821720 |
BC003004.1 |
- |
4, 5 |
362 |
2821720 |
BC107697.1 |
- |
4, 5 |
380 |
6454606 |
BG715167.1 |
- |
4, 5 |
392 |
|
BI596981.1 |
- |
4, 5 |
364 |
|
CN257020.1 |
- |
2, 3 |
163 |
|
CR541889.1 |
- |
4, 5 |
352 |
|
View Full Product Details