Gene Transcripts

Gene Symbol
Myl6
Entrez Gene ID
685867
Gene Name
myosin, light chain 6, alkali, smooth muscle and non-muscle
Gene Aliases
Chromosome Location
Chr. 7: 2906137 - 2909179 on Build Rnor_6.0
UniGene ID
Rn.146093
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109484.1 NP_001102954.1 6 533
XM_008765039.1 XP_008763261.1 5 1559
XM_008765040.1 XP_008763262.1 7 572
GenBank mRNA BC166586.1 - 5 1530
FQ217612.1 - 6 494
FQ217952.1 - 6 506
FQ221192.1 - 6 500
FQ221499.1 - 6 502
FQ221709.1 - 6 534
FQ221718.1 - 6 505
FQ221751.1 - 6 501
FQ221963.1 - 6 502
FQ222034.1 - 6 493
FQ222160.1 - 6 503
FQ222384.1 - 6 505
FQ222504.1 - 6 482
FQ223317.1 - 6 508
FQ228533.1 - 6 505

Gene Symbol
Myl6b
Entrez Gene ID
685883
Gene Name
myosin, light chain 6B, alkali, smooth muscle and non-muscle
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.146093
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109486.1 NP_001102956.1 7 478

Gene Symbol
Myl6l
Entrez Gene ID
362816
Gene Name
myosin, light polypeptide 6, alkali, smooth muscle and non-muscle-like
Gene Aliases
LC17, MLC-3, Myl6
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.146093
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100983.1 NP_001094453.1 5 477
GenBank mRNA S77858.1 - 5 477

View Full Product Details