Gene Transcripts
- Gene Symbol
- Bcl2a1d
- Entrez Gene ID
- 12047
- Gene Name
- B cell leukemia/lymphoma 2 related protein A1d
- Gene Aliases
- A1-d
- Chromosome Location
- Chr. 9: 88723288 - 88731850 on Build GRCm38
- UniGene ID
- Mm.378888
- Species
- Mus musculus
- Gene Symbol
- Bcl2a1a
- Entrez Gene ID
- 12044
- Gene Name
- B cell leukemia/lymphoma 2 related protein A1a
- Gene Aliases
- A1, BB218357, Bcl2a1, Bfl-1, Hbpa1
- Chromosome Location
- Chr. 9: 88956920 - 88962416 on Build GRCm38
- UniGene ID
- Mm.425593
- Species
- Mus musculus
- Gene Symbol
- Bcl2a1b
- Entrez Gene ID
- 12045
- Gene Name
- B cell leukemia/lymphoma 2 related protein A1b
- Gene Aliases
- A1-b
- Chromosome Location
- Chr. 9: 89199273 - 89207838 on Build GRCm38
- UniGene ID
- Mm.479217
- Species
- Mus musculus
View Full Product Details