Gene Transcripts

Gene Symbol
RBPJL
Entrez Gene ID
11317
Gene Name
recombination signal binding protein for immunoglobulin kappa J region like
Gene Aliases
RBPL, RBPSUHL, SUHL
Chromosome Location
Chr. 20: 45306836 - 45319677 on Build GRCh38
UniGene ID
Hs.248217
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014276.3 NP_055091.2 11, 12 1352
XM_011528522.2 XP_011526824.1 11, 12 1264

View Full Product Details