Gene Transcripts

Gene Symbol
RAB40B
Entrez Gene ID
10966
Gene Name
RAB40B, member RAS oncogene family
Gene Aliases
RAR, SEC4L
Chromosome Location
Chr. 17: 82657067 - 82698722 on Build GRCh38
UniGene ID
Hs.484068
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006822.2 NP_006813.1 6 1234
XM_005256334.4 XP_005256391.1 6 1437
XM_006722271.3 XP_006722334.1 6 1003
XM_011523528.2 XP_011521830.1 6 1356
XM_017024042.1 XP_016879531.1 7 1869
XM_017024043.1 XP_016879532.1 6 1346
GenBank mRNA AK095782.1 - 1 1998
AK124376.1 - 2 1850
BC018039.1 - 6 1234 4801271
U05227.1 - 6 1153

View Full Product Details