Gene Transcripts

Gene Symbol
MGAT4A
Entrez Gene ID
11320
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme A
Gene Aliases
GNT-IV, GNT-IVA, GnT-4a
Chromosome Location
Chr. 2: 98619106 - 98731126 on Build GRCh38
UniGene ID
Hs.177576
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160154.1 NP_001153626.1 10 1070
NM_012214.2 NP_036346.1 13 1654
GenBank mRNA AB000616.1 - 12 1475
AK291533.1 - 12 1464
AK304393.1 - 10 1070
AK314024.1 - 13 1614
BC127107.1 - 12 1552 40124731
BC127108.1 - 12 1552 40124741

View Full Product Details