Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001105571.2 | NP_001099041.1 | 1 | 128 | |
NM_001220493.1 | NP_001207422.1 | 1 | 128 | ||
GenBank mRNA | BC147024.1 | - | 1 | 128 | |
BC147025.1 | - | 1 | 128 | 9056953 | |
BC171909.1 | - | 1 | 128 | 9054563 |