Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005546.3 | NP_005537.3 | 6, 7 | 731 | |
XM_017009443.1 | XP_016864932.1 | 4, 5 | 344 | ||
GenBank mRNA | AB209622.1 | - | 6, 7 | 784 | |
AK312846.1 | - | 6, 7 | 700 | ||
BC109077.2 | - | 6, 7 | 722 | 40034274 | |
BC109078.1 | - | 6, 7 | 722 | 40034276 | |
D13720.1 | - | 6, 7 | 731 |