Gene Transcripts

Gene Symbol
MARCH4
Entrez Gene ID
57574
Gene Name
membrane associated ring-CH-type finger 4
Gene Aliases
MARCH-IV, RNF174
Chromosome Location
Chr. 2: 216257862 - 216372027 on Build GRCh38
UniGene ID
Hs.170388
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020814.2 NP_065865.1 3 2536
GenBank mRNA AB037820.1 - 3 2536
BC048793.1 - 3 610
BC098448.1 - 3 965 6209260

View Full Product Details