Gene Transcripts
- Gene Symbol
- SSX1
- Entrez Gene ID
- 6756
- Gene Name
- SSX family member 1
- Gene Aliases
- CT5.1, SSRC
- Chromosome Location
- Chr. X: 48255317 - 48267444 on Build GRCh38
- UniGene ID
- Hs.434142
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278691.1 |
NP_001265620.1
|
5, 6 |
499 |
|
NM_005635.3 |
NP_005626.1
|
4, 5 |
413 |
|
GenBank mRNA |
BC001003.2 |
- |
4, 5 |
358 |
|
BC125151.1 |
- |
4, 5 |
356 |
40120554 |
BC128611.1 |
- |
4, 5 |
355 |
6159943 |
BC133693.1 |
- |
4, 5 |
356 |
40120550 |
BC150487.2 |
- |
4, 5 |
360 |
40120555 |
BE777476.1 |
- |
5, 6 |
433 |
|
BF211314.1 |
- |
4, 5 |
378 |
|
X86174.1 |
- |
4, 5 |
368 |
|
View Full Product Details