Gene Transcripts

Gene Symbol
PIP4K2B
Entrez Gene ID
8396
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 beta
Gene Aliases
PI5P4KB, PIP5K2B, PIP5KIIB, PIP5KIIbeta, PIP5P4KB
Chromosome Location
Chr. 17: 38765691 - 38799965 on Build GRCh38
UniGene ID
Hs.260603
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003559.4 NP_003550.1 5 1079
XM_011525326.2 XP_011523628.1 6 1220
XM_011525327.2 XP_011523629.1 7 739
XM_011525329.2 XP_011523631.1 6 1220
XM_011525330.2 XP_011523632.1 4 415
XM_017025196.1 XP_016880685.1 6 656
XM_017025197.1 XP_016880686.1 6 556
XM_017025198.1 XP_016880687.1 4 1042
XM_017025199.1 XP_016880688.1 5 494
GenBank mRNA BC027459.1 - 5 696
BT019614.1 - 5 598
U85245.1 - 5 1078

View Full Product Details