Gene Transcripts

Gene Symbol
TYW5
Entrez Gene ID
129450
Gene Name
tRNA-yW synthesizing protein 5
Gene Aliases
C2orf60, hTYW5
Chromosome Location
Chr. 2: 199928911 - 199955736 on Build GRCh38
UniGene ID
Hs.204619
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039693.2 NP_001034782.1 8 1111
NR_004862.1 8 1092
NR_109905.1 5 840
NR_109906.1 7 1004
NR_109907.1 7 956
XM_017003310.1 XP_016858799.1 6 739
GenBank mRNA AK095272.1 - 7 720
AK293064.1 - 7 771
AK298977.1 - 8 855
BC025758.1 - 7 751
BC063502.1 - 4 380
BC132835.1 - 8 875 40146857
BC136837.1 - 8 875 9020828
BC144420.1 - 8 856
BX647677.1 - 8 1094

View Full Product Details