Gene Transcripts
- Gene Symbol
- GYPB
- Entrez Gene ID
- 2994
- Gene Name
- glycophorin B (MNS blood group)
- Gene Aliases
- CD235b, GPB, GYP, MNS, PAS-3, SS
- Chromosome Location
- Chr. 4: 143995172 - 144021860 on Build GRCh38
- UniGene ID
- Hs.654368
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304382.1 |
NP_001291311.1
|
4, 5 |
305 |
|
NM_002100.5 |
NP_002091.3
|
3, 4 |
225 |
|
XM_011531903.2 |
XP_011530205.1
|
3, 4 |
360 |
|
XM_011531904.2 |
XP_011530206.1
|
4, 5 |
2864 |
|
XM_011531905.2 |
XP_011530207.1
|
4, 5 |
512 |
|
XM_017008137.1 |
XP_016863626.1
|
3, 4 |
2559 |
|
GenBank mRNA |
AY950609.1 |
- |
3, 4 |
167 |
|
AY950612.1 |
- |
4, 5 |
260 |
|
BC069310.1 |
- |
3, 4 |
217 |
7262104 |
BC121077.1 |
- |
3, 4 |
201 |
40119713 |
BC121078.1 |
- |
3, 4 |
201 |
40119716 |
EU338223.1 |
- |
2, 3 |
105 |
|
EU338224.1 |
- |
4, 5 |
328 |
|
EU338225.1 |
- |
4, 5 |
297 |
|
EU338226.1 |
- |
4, 5 |
262 |
|
EU338228.1 |
- |
2, 3 |
105 |
|
EU338229.1 |
- |
4, 5 |
328 |
|
EU338235.1 |
- |
2, 3 |
105 |
|
EU338236.1 |
- |
4, 5 |
328 |
|
EU338237.1 |
- |
5, 6 |
421 |
|
J02982.1 |
- |
3, 4 |
177 |
|
X08055.1 |
- |
5, 6 |
303 |
|
View Full Product Details