Gene Transcripts

Gene Symbol
WDR47
Entrez Gene ID
22911
Gene Name
WD repeat domain 47
Gene Aliases
Chromosome Location
Chr. 1: 108970211 - 109042228 on Build GRCh38
UniGene ID
Hs.570055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142550.1 NP_001136022.1 3 546
NM_001142551.1 NP_001136023.1 3 546
NM_014969.5 NP_055784.3 3 546
XM_006710460.3 XP_006710523.1 3 538
XM_011541027.2 XP_011539329.1 3 538
XM_011541028.2 XP_011539330.1 3 538
XM_011541029.1 XP_011539331.1 3 538
XM_011541030.1 XP_011539332.1 3 207
XM_011541031.1 XP_011539333.1 1 12
XM_017000696.1 XP_016856185.1 3 538
XM_017000697.1 XP_016856186.1 3 207
GenBank mRNA AB020700.1 - 3 393
AK225781.1 - 3 305
AK289789.1 - 3 420
BC034964.2 - 3 431 4823902

View Full Product Details