Gene Transcripts

Gene Symbol
FBXW5
Entrez Gene ID
54461
Gene Name
F-box and WD repeat domain containing 5
Gene Aliases
Fbw5
Chromosome Location
Chr. 9: 136940433 - 136944774 on Build GRCh38
UniGene ID
Hs.522507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018998.3 NP_061871.1 6 895
XM_005266089.2 XP_005266146.2 6 945
XM_011518789.1 XP_011517091.1 6 945
XM_017014812.1 XP_016870301.1 6 915
GenBank mRNA AB209726.1 - 3 307
AF217998.1 - 2 57
AF258569.1 - 2 57
AK026081.1 - 6 759
AK315738.1 - 6 770
AL137631.1 - 2 158
BC004541.2 - 3 304
BC009429.1 - 1 48
BC014130.1 - 6 791 4620371
BC014297.2 - 6 762 3958395
BC031100.1 - 5 910
BC035355.1 - 5 909

View Full Product Details