Gene Transcripts
- Gene Symbol
- TRAPPC11
- Entrez Gene ID
- 60684
- Gene Name
- trafficking protein particle complex 11
- Gene Aliases
- C4orf41, FOIGR, GRY, LGMD2S
- Chromosome Location
- Chr. 4: 183659267 - 183713594 on Build GRCh38
- UniGene ID
- Hs.443240
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021942.5 |
NP_068761.4
|
28, 29 |
3389 |
|
NM_199053.2 |
NP_951008.1
|
28, 29 |
3389 |
|
XM_017008539.1 |
XP_016864028.1
|
26, 27 |
3040 |
|
GenBank mRNA |
AK022778.1 |
- |
7, 8 |
793 |
|
AK023390.1 |
- |
9, 10 |
1078 |
|
AK090489.1 |
- |
3, 4 |
1180 |
|
AK091146.1 |
- |
23, 24 |
2613 |
|
AK096345.1 |
- |
28, 29 |
3363 |
|
AK315057.1 |
- |
23, 24 |
2595 |
|
AL136752.1 |
- |
20, 21 |
2298 |
|
AL833451.1 |
- |
27, 28 |
3195 |
|
AL833571.1 |
- |
29, 30 |
3309 |
|
BC051724.1 |
- |
28, 29 |
3333 |
5265781 |
BC139745.1 |
- |
16, 17 |
1769 |
|
BX538044.1 |
- |
28, 29 |
3343 |
|
BX647127.1 |
- |
28, 29 |
3360 |
|
BX648499.1 |
- |
28, 29 |
3364 |
|
CR533553.1 |
- |
20, 21 |
2233 |
|
View Full Product Details