Gene Transcripts

Gene Symbol
RPS21
Entrez Gene ID
6227
Gene Name
ribosomal protein S21
Gene Aliases
HLDF, S21
Chromosome Location
Chr. 20: 62387065 - 62388520 on Build GRCh38
UniGene ID
Hs.190968
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024.3 NP_001015.1 4, 5 277
GenBank mRNA AV758927.1 - 238
BC018140.1 - 3, 4 214 3903320
BC021990.1 - 4, 5 242 4066330
BM840443.1 - 4, 5 277
CF124568.1 - 4, 5 318
CR542132.1 - 3, 4 187
L04483.1 - 4, 5 225
X79563.2 - 4, 5 223

View Full Product Details