Gene Transcripts

Gene Symbol
MT1IP
Entrez Gene ID
644314
Gene Name
metallothionein 1I, pseudogene
Gene Aliases
MT1, MT1I, MTE
Chromosome Location
Chr. 16: 56676116 - 56677877 on Build GRCh38
UniGene ID
Hs.434199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003669.2 3 274
NR_104045.1 3 259
NR_104046.1 2 230
GenBank mRNA AF348997.1 - 3 297

View Full Product Details